Product Details

SNP ID
rs2131237
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:40426274 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAGGGAGAGAAAGGCAGAGGGGC[A/G]GGCAAACAGCAATTATTGTAAGCAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RBM47 PubMed Links

Gene Details

Gene
RBM47
Gene Name
RNA binding motif protein 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098634.1 Intron NP_001092104.1
NM_019027.3 Intron NP_061900.2
XM_005248103.3 Intron XP_005248160.1
XM_005248107.3 Intron XP_005248164.1
XM_005248108.3 Intron XP_005248165.1
XM_005248109.4 Intron XP_005248166.1
XM_011513704.2 Intron XP_011512006.1
XM_011513707.2 Intron XP_011512009.1
XM_011513708.2 Intron XP_011512010.1
XM_017008304.1 Intron XP_016863793.1
XM_017008305.1 Intron XP_016863794.1
XM_017008306.1 Intron XP_016863795.1
XM_017008307.1 Intron XP_016863796.1
XM_017008308.1 Intron XP_016863797.1
XM_017008309.1 Intron XP_016863798.1
XM_017008310.1 Intron XP_016863799.1

View Full Product Details