Product Details

SNP ID
rs526123
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:46856542 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGACAGAATGACTTGAGAAGCAAA[A/G]ACAATGGAATGATCTATGCCAAATA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADGRF5 PubMed Links

Gene Details

Gene
ADGRF5
Gene Name
adhesion G protein-coupled receptor F5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098518.1 Intron NP_001091988.1
NM_015234.4 Intron NP_056049.4
XM_005248892.2 Intron XP_005248949.1
XM_005248893.3 Intron XP_005248950.1
XM_005248894.3 Intron XP_005248951.1
XM_005248895.3 Intron XP_005248952.1
XM_011514353.2 Intron XP_011512655.1
XM_011514354.2 Intron XP_011512656.1
XM_017010418.1 Intron XP_016865907.1
XM_017010419.1 Intron XP_016865908.1
XM_017010420.1 Intron XP_016865909.1
XM_017010421.1 Intron XP_016865910.1
XM_017010422.1 Intron XP_016865911.1
XM_017010423.1 Intron XP_016865912.1
XM_017010424.1 Intron XP_016865913.1

View Full Product Details