Product Details

SNP ID
rs2073768
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:19788496 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAACGTCTCCTGCAGGCCTCGGAC[A/G]GCCAGGGCTCTGGCTGGCCCCCAGA
Phenotype
MIM: 610778 MIM: 602054
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GNB1L PubMed Links

Gene Details

Gene
GNB1L
Gene Name
G protein subunit beta 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_053004.2 1434 UTR 3 NP_443730.1
Gene
TBX1
Gene Name
T-box 1
There are no transcripts associated with this gene.

View Full Product Details