Product Details

SNP ID
rs2285835
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:20855239 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTATATCTGTGGGACCAATTCATC[C/T]AAGCCCCATGTGCCTATTGCTTCAC
Phenotype
MIM: 616167
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCUN1D3 PubMed Links

Gene Details

Gene
DCUN1D3
Gene Name
defective in cullin neddylation 1 domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173475.3 5822 UTR 3 NP_775746.1
Gene
LOC81691
Gene Name
exonuclease NEF-sp
There are no transcripts associated with this gene.

View Full Product Details