Product Details

SNP ID
rs2273607
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:100732089 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCCTGTGCCAACAACAGGACCTGC[A/G]TGAGCCTGGACGATGGCCTCTATGA
Phenotype
MIM: 176290
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DLK1 PubMed Links

Gene Details

Gene
DLK1
Gene Name
delta like non-canonical Notch ligand 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317172.1 514 Missense Mutation ATG,GTG M104V NP_001304101.1
NM_003836.6 514 Missense Mutation ATG,GTG M104V NP_003827.3

View Full Product Details