Product Details

SNP ID
rs2298552
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60497474 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGTGCTCAGCGTGCTCAGCCCTA[C/T]GGCATCACATCTCCGGGAATCTTTG
Phenotype
MIM: 606550
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MS4A12 PubMed Links

Gene Details

Gene
MS4A12
Gene Name
membrane spanning 4-domains A12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164470.1 213 Silent Mutation TAC,TAT Y52Y NP_001157942.1
NM_017716.2 213 Silent Mutation TAC,TAT Y52Y NP_060186.2
XM_011545117.2 213 Nonsense Mutation TAC,TAT Y52Y XP_011543419.1

View Full Product Details