Product Details
- SNP ID
-
rs2503129
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:71153102 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTCTCCAGCTCGACCTGCCTGTGT[A/T]CACACAAGCAGCCCCTTCCACCCAG
- Phenotype
-
MIM: 300336
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NLGN3
PubMed Links
Gene Details
- Gene
- NLGN3
- Gene Name
- neuroligin 3
There are no transcripts associated with this gene.
View Full Product Details