Product Details

SNP ID
rs74572140
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:148865715 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGCCAATACTTCACCTGTTCTGG[G/T]TTTGGTCTTGGTTTCCAAGACTACC
Phenotype
MIM: 114851 MIM: 114852
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CPA3 PubMed Links

Gene Details

Gene
CPA3
Gene Name
carboxypeptidase A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001870.3 Intron NP_001861.2
Gene
CPB1
Gene Name
carboxypeptidase B1
There are no transcripts associated with this gene.

View Full Product Details