Product Details
- SNP ID
-
rs7216358
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:70174878 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTAAAGTAACTCTGCATGTCAGTAG[A/T]CAGACCTTGGTAGAACCACAAGGCT
- Phenotype
-
MIM: 600681
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
KCNJ2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs536993975] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KCNJ2
- Gene Name
- potassium voltage-gated channel subfamily J member 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000891.2 |
222 |
UTR 5 |
|
|
NP_000882.1 |
- Gene
- KCNJ2-AS1
- Gene Name
- KCNJ2 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.
View Full Product Details