Product Details

SNP ID
rs61754163
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53211712 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTCAGGCCCCCTTTGCCCTGGCC[C/T]CCAGGAACCTCATCCTCGCTAGAGG
Phenotype
MIM: 147559 MIM: 180190
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ITGB7 PubMed Links

Gene Details

Gene
ITGB7
Gene Name
integrin subunit beta 7
There are no transcripts associated with this gene.

Gene
RARG
Gene Name
retinoic acid receptor gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000966.5 1495 Silent Mutation GGA,GGG G443G NP_000957.1
NM_001042728.2 1495 Silent Mutation GGA,GGG G432G NP_001036193.1
NM_001243730.1 1495 Silent Mutation GGA,GGG G371G NP_001230659.1
NM_001243731.1 1495 Silent Mutation GGA,GGG G322G NP_001230660.1
NM_001243732.1 1495 Silent Mutation GGA,GGG G421G NP_001230661.1

View Full Product Details