Product Details
- SNP ID
-
rs368003231
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:11436245 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCCAGGCTGGAGGTGGGAGGGGCC[A/G]ACATTGGGCCTTAGGGATAGGCATT
- Phenotype
-
MIM: 615956
MIM: 177060
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC151
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs143936796,rs146514973] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCDC151
- Gene Name
- coiled-coil domain containing 151
- Gene
- PRKCSH
- Gene Name
- protein kinase C substrate 80K-H
View Full Product Details