Product Details

SNP ID
rs12720063
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:219574956 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGCCATGCTCCCCCTCACTGGGC[C/T]GTGCTGCACCTGGCCACCTCTGCTC
Phenotype
MIM: 147380 MIM: 610991
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
INHA PubMed Links

Gene Details

Gene
INHA
Gene Name
inhibin alpha subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002191.3 674 Silent Mutation GCC,GCT A177A NP_002182.1
Gene
OBSL1
Gene Name
obscurin like 1
There are no transcripts associated with this gene.

View Full Product Details