Product Details

SNP ID
rs59820101
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81245652 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCTGGCGGAGCTGGGCATCCAG[C/G]GCGCCACGGAGGTCGTTCACCTGGA
Phenotype
MIM: 616525
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C17orf89 PubMed Links

Gene Details

Gene
C17orf89
Gene Name
chromosome 17 open reading frame 89
There are no transcripts associated with this gene.

Gene
SLC38A10
Gene Name
solute carrier family 38 member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037984.2 3076 Silent Mutation GCC,GCG A1088A NP_001033073.1
NM_138570.3 3076 Intron NP_612637.1
XM_005257019.1 3076 Silent Mutation GCC,GCG A1096A XP_005257076.1
XM_011524288.1 3076 Silent Mutation GCC,GCG A1103A XP_011522590.1
XM_011524289.1 3076 Silent Mutation GCC,GCG A1095A XP_011522591.1
XM_011524290.1 3076 Silent Mutation GCC,GCG A1014A XP_011522592.1
Gene
TEPSIN
Gene Name
TEPSIN, adaptor related protein complex 4 accessory protein
There are no transcripts associated with this gene.

View Full Product Details