Product Details

SNP ID
rs61740674
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56441363 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTCCAGTATGACTTCTTTCATG[C/T]AGAATAAGAGATGTGCGCTGACTAA
Phenotype
MIM: 611024
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF667 PubMed Links

Gene Details

Gene
ZNF667
Gene Name
zinc finger protein 667
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321355.1 1821 Silent Mutation CTA,CTG L487L NP_001308284.1
NM_001321356.1 1821 Silent Mutation CTA,CTG L544L NP_001308285.1
NM_022103.3 1821 Silent Mutation CTA,CTG L544L NP_071386.3
XM_011527208.1 1821 Silent Mutation CTA,CTG L544L XP_011525510.1
XM_011527209.1 1821 Silent Mutation CTA,CTG L544L XP_011525511.1

View Full Product Details