Product Details

SNP ID
rs58774681
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:40525900 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTTTCAGTTTTTTTGTTTGTTT[G/T]TTTATGTTTTTTTACTCACATTAGC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF620 PubMed Links

Gene Details

Gene
ZNF620
Gene Name
zinc finger protein 620
There are no transcripts associated with this gene.

Gene
ZNF621
Gene Name
zinc finger protein 621
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098414.2 Intron NP_001091884.1
NM_001287245.1 Intron NP_001274174.1
NM_198484.4 Intron NP_940886.1
XM_005265079.3 Intron XP_005265136.1
XM_005265080.3 Intron XP_005265137.1
XM_006713114.3 Intron XP_006713177.1
XM_011533610.2 Intron XP_011531912.1
XM_017006226.1 Intron XP_016861715.1

View Full Product Details