Product Details

SNP ID
rs3087367
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:44116547 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCAGAAGGCCATCAGGCCCAGGC[A/G]AGTCCCAGGCTCAGAGGAGAGTAGA
Phenotype
MIM: 602981 MIM: 600815
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AEBP1 PubMed Links

Gene Details

Gene
AEBP1
Gene Name
AE binding protein 1
There are no transcripts associated with this gene.

Gene
MIR4649
Gene Name
microRNA 4649
There are no transcripts associated with this gene.

Gene
POLD2
Gene Name
DAN polymerase delta 2, accessory subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127218.2 Intron NP_001120690.1
NM_001256879.1 Intron NP_001243808.1
NM_006230.3 Intron NP_006221.2
XM_011515431.1 Intron XP_011513733.1

View Full Product Details