Product Details

SNP ID
rs201554877
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:43609361 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCACACATGCAGGTGAGTGGGT[A/G]GGGAACATCCCAGCAGAAGCTGAGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF485 PubMed Links

Gene Details

Gene
ZNF485
Gene Name
zinc finger protein 485
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318140.1 Intron NP_001305069.1
NM_001318141.1 Intron NP_001305070.1
NM_001318142.1 Intron NP_001305071.1
NM_001318143.1 Intron NP_001305072.1
NM_145312.3 Intron NP_660355.2
XM_011539498.2 Intron XP_011537800.1

View Full Product Details