Product Details

SNP ID
rs57144070
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:46350970 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGGAGGGTGGAAGGTGGGGGCA[C/T]TAGGTACACTCAGGTGGGCAGATCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC35C2 PubMed Links

Gene Details

Gene
SLC35C2
Gene Name
solute carrier family 35 member C2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281457.1 Intron NP_001268386.1
NM_001281458.1 Intron NP_001268387.1
NM_001281459.1 Intron NP_001268388.1
NM_001281460.1 Intron NP_001268389.1
NM_015945.11 Intron NP_057029.8
NM_173073.3 Intron NP_775096.1
NM_173179.3 Intron NP_775271.1
XM_011528831.1 Intron XP_011527133.1
XM_011528832.1 Intron XP_011527134.1
XM_011528833.1 Intron XP_011527135.1
XM_011528834.1 Intron XP_011527136.1
XM_011528835.1 Intron XP_011527137.1
XM_011528836.1 Intron XP_011527138.1
XM_011528837.1 Intron XP_011527139.1
XM_011528838.1 Intron XP_011527140.1
XM_017027856.1 Intron XP_016883345.1
XM_017027857.1 Intron XP_016883346.1
XM_017027858.1 Intron XP_016883347.1
XM_017027859.1 Intron XP_016883348.1
XM_017027860.1 Intron XP_016883349.1
XM_017027861.1 Intron XP_016883350.1
XM_017027862.1 Intron XP_016883351.1

View Full Product Details