Product Details

SNP ID
rs28592687
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:31704398 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTCCACGTGGTGGAAGGTTGGTC[T/C]GGAAGTTGGGCAGCCTGCCCAGGGA
Phenotype
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
CLUHP3 PubMed Links

Gene Details

Gene
CLUHP3
Gene Name
clustered mitochondria homolog pseudogene 3
There are no transcripts associated with this gene.

Gene
ZNF720
Gene Name
zinc finger protein 720
There are no transcripts associated with this gene.

View Full Product Details