Product Details

SNP ID
rs74075111
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:27081945 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGCACTTGGGCAGGTCTTGACAA[A/G]CATCATATTCCTGATGACTGACCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C12orf71 PubMed Links

Gene Details

Gene
C12orf71
Gene Name
chromosome 12 open reading frame 71
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080406.1 Intron NP_001073875.1
XM_011520847.2 Intron XP_011519149.1

View Full Product Details