Product Details

SNP ID
rs74086001
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:103532573 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGATGCCTTGCCCATCCCTTCTTG[A/T]CCTGCCAGGCACCGGCAGTGGCTCT
Phenotype
MIM: 123280
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CKB PubMed Links

Gene Details

Gene
CKB
Gene Name
creatine kinase B
There are no transcripts associated with this gene.

Gene
TRMT61A
Gene Name
tRNA methyltransferase 61A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152307.2 Intron NP_689520.2

View Full Product Details