Product Details

SNP ID
rs61750906
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:36170583 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCCCAACCCAGAAGTGCAGTGC[C/T]GAGGGAGAAGCTTTGAGACCGGGGG
Phenotype
MIM: 616066
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC169-SOHLH2 PubMed Links

Gene Details

Gene
CCDC169-SOHLH2
Gene Name
CCDC169-SOHLH2 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198910.1 1751 Missense Mutation CAG,CGG Q479R NP_001185839.1
Gene
SOHLH2
Gene Name
spermatogenesis and oogenesis specific basic helix-loop-helix 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282147.1 1751 Intron NP_001269076.1
NM_017826.2 1751 Missense Mutation CAG,CGG Q402R NP_060296.2

View Full Product Details