Product Details
- SNP ID
-
rs61750912
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:12096424 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGTGGGGACCCACGCGGGTTCAAC[A/G]TGCATATCGAGAAGTGTTATTTCTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF788
PubMed Links
Gene Details
- Gene
- ZNF788
- Gene Name
- zinc finger family member 788
There are no transcripts associated with this gene.
View Full Product Details