Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001319109.1 | 1538 | UTR 3 | NP_001306038.1 | ||
NM_001319110.1 | 1538 | UTR 3 | NP_001306039.1 | ||
NM_001319111.1 | 1538 | UTR 3 | NP_001306040.1 | ||
NM_001319112.1 | 1538 | UTR 3 | NP_001306041.1 | ||
NM_001319114.1 | 1538 | UTR 3 | NP_001306043.1 | ||
NM_001319115.1 | 1538 | UTR 3 | NP_001306044.1 | ||
NM_015412.3 | 1538 | UTR 3 | NP_056227.2 | ||
XM_005247276.2 | 1538 | UTR 3 | XP_005247333.1 | ||
XM_011512635.2 | 1538 | UTR 3 | XP_011510937.1 | ||
XM_011512636.2 | 1538 | UTR 3 | XP_011510938.1 | ||
XM_011512638.2 | 1538 | UTR 3 | XP_011510940.1 | ||
XM_017006102.1 | 1538 | UTR 3 | XP_016861591.1 | ||
XM_017006103.1 | 1538 | Intron | XP_016861592.1 |