Product Details

SNP ID
rs113127192
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:3577803 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTATTTTAGTAGTTTCAGAAATGT[A/G]TGTACCCCTCTTATTAACAACTCTT
Phenotype
MIM: 603658
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPS7 PubMed Links

Gene Details

Gene
RPS7
Gene Name
ribosomal protein S7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011.3 Intron NP_001002.1

View Full Product Details