Product Details
- SNP ID
-
hCV25926487
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:136226151 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACCTTTTGAATGAGGGTTCCTATA[C/T]CATGATATTTGCGGATTGTCAATTT
- Phenotype
-
MIM: 300930
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MAP7D3
PubMed Links
Gene Details
- Gene
- MAP7D3
- Gene Name
- MAP7 domain containing 3
View Full Product Details