Product Details

SNP ID
rs112933980
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:138558673 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAGAATACCTAGGGAAGAAGAAA[A/C]CCTCCTGGGTTGTCAATGTGATTAA
Phenotype
MIM: 600548
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HSPA9 PubMed Links

Gene Details

Gene
HSPA9
Gene Name
heat shock protein family A (Hsp70) member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004134.6 Intron NP_004125.3
Gene
SNORD63
Gene Name
small nucleolar RNA, C/D box 63
There are no transcripts associated with this gene.

View Full Product Details