Product Details

SNP ID
hCV25972713
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:1309344 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATCTGTCGCGGGCGGAGGCTGGA[G/A]ATCTGGACAGACTTCCTTGTCTGGT
Phenotype
MIM: 611354
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
ACAP3 PubMed Links

Gene Details

Gene
ACAP3
Gene Name
ArfGAP with coiled-coil, ankyrin repeat and PH domains 3
There are no transcripts associated with this gene.

Gene
CPSF3L
Gene Name
cleavage and polyadenylation specific factor 3-like
There are no transcripts associated with this gene.

Gene
MIR6727
Gene Name
microRNA 6727
There are no transcripts associated with this gene.

Gene
PUSL1
Gene Name
pseudouridylate synthase-like 1
There are no transcripts associated with this gene.

View Full Product Details