Product Details

SNP ID
rs8282
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45252257 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGATGTGCAAAGAGAAATAGGCT[C/T]GTTTATTTATTCATTGATCAACTGG
Phenotype
MIM: 107285
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLPI PubMed Links

Gene Details

Gene
SLPI
Gene Name
secretory leukocyte peptidase inhibitor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003064.3 578 UTR 3 NP_003055.1

View Full Product Details