Product Details

SNP ID
rs9988418
Assay Type
Functionally tested
NCBI dbSNP Submissions
51
Location
Chr.1:18839606 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGACGCCACCGGATGGCACTAGTCC[C/T]TCCTCATGGTGTAGCCCTGGATCAT
Phenotype
MIM: 606226
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TAS1R2 PubMed Links

Gene Details

Gene
TAS1R2
Gene Name
taste 1 receptor member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152232.2 2514 Missense Mutation AAG,AGG K838R NP_689418.2

View Full Product Details