Product Details

SNP ID
rs56015130
Assay Type
Functionally tested
NCBI dbSNP Submissions
45
Location
Chr.1:27668566 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGAGCAGACAAAGCGTAGTGGGG[G/T]TGTTGGGGTGGGACACAGGGGTCCC
Phenotype
MIM: 147572
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IFI6 PubMed Links

Gene Details

Gene
IFI6
Gene Name
interferon alpha inducible protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002038.3 Intron NP_002029.3
NM_022872.2 Intron NP_075010.1
NM_022873.2 Intron NP_075011.1

View Full Product Details