Product Details

SNP ID
rs28576169
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:28495561 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGTGAACAGAGAAGAGAGGCTG[A/G]GAAGCTGGGAACAGGAGGAGGAGGA
Phenotype
MIM: 605220 MIM: 607042 MIM: 608273
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APOBR PubMed Links

Gene Details

Gene
APOBR
Gene Name
apolipoprotein B receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018690.3 553 Missense Mutation AGA,GGA R174G NP_061160.3
Gene
CLN3
Gene Name
CLN3, battenin
There are no transcripts associated with this gene.

Gene
IL27
Gene Name
interleukin 27
There are no transcripts associated with this gene.

View Full Product Details