Product Details

SNP ID
rs116301817
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154398354 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCGCAGACAGACCGCCTATATAA[A/G]CCATGCGCAGGCGGAGGAGCGCCTC
Phenotype
MIM: 300081 MIM: 312173
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNASE1L1 PubMed Links

Gene Details

Gene
DNASE1L1
Gene Name
deoxyribonuclease I-like 1
There are no transcripts associated with this gene.

Gene
LOC107987332
Gene Name
uncharacterized LOC107987332
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017030021.1 290 Intron XP_016885510.1
Gene
RPL10
Gene Name
ribosomal protein L10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256577.2 290 UTR 5 NP_001243506.2
NM_001256580.2 290 UTR 5 NP_001243509.2
NM_001303624.1 290 Intron NP_001290553.1
NM_001303625.1 290 UTR 5 NP_001290554.1
NM_001303626.1 290 Intron NP_001290555.1
NM_006013.4 290 UTR 5 NP_006004.3
Gene
SNORA70
Gene Name
small nucleolar RNA, H/ACA box 70
There are no transcripts associated with this gene.

View Full Product Details