Product Details
- SNP ID
-
hCV25997775
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:66964259 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGACAGGCCAGACCTGGGGAGCA[A/G]AGAAGGGTCTGGCATCCTGGTGGCT
- Phenotype
-
MIM: 605279
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CES3
PubMed Links
Gene Details
- Gene
- CES3
- Gene Name
- carboxylesterase 3
View Full Product Details