Product Details

SNP ID
rs2282200
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:92147394 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACTGATGTAACAACCTTGGTGTGC[C/T]GTGGCTAAGCCACACTCCACCCCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00475 PubMed Links

Gene Details

Gene
LINC00475
Gene Name
long intergenic non-protein coding RNA 475
There are no transcripts associated with this gene.

Gene
LOC100128076
Gene Name
protein tyrosine phosphatase pseudogene
There are no transcripts associated with this gene.

View Full Product Details