Product Details

SNP ID
rs11942947
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:174292780 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTATTGTTTTCCTGATTTTTATTTC[A/G]TTTTCTATCTGGGTTTGTTTATTTG
Phenotype
MIM: 605649
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEP44 PubMed Links

Gene Details

Gene
CEP44
Gene Name
centrosomal protein 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040157.2 Intron NP_001035247.1
NM_001145314.1 Intron NP_001138786.1
XM_011532285.2 Intron XP_011530587.1
XM_011532286.2 Intron XP_011530588.1
XM_011532287.2 Intron XP_011530589.1
XM_017008654.1 Intron XP_016864143.1
XM_017008655.1 Intron XP_016864144.1
XM_017008656.1 Intron XP_016864145.1
XM_017008657.1 Intron XP_016864146.1
XM_017008658.1 Intron XP_016864147.1
XM_017008659.1 Intron XP_016864148.1
XM_017008660.1 Intron XP_016864149.1
Gene
FBXO8
Gene Name
F-box protein 8
There are no transcripts associated with this gene.

View Full Product Details