Product Details

SNP ID
rs13049218
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:36159592 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAATGACGATTTCCAAACTTCTCT[C/T]GTGTGCTGGTAGTTACACCAAATAG
Phenotype
MIM: 604803
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CBR3-AS1 PubMed Links

Gene Details

Gene
CBR3-AS1
Gene Name
CBR3 antisense RNA 1
There are no transcripts associated with this gene.

Gene
DOPEY2
Gene Name
dopey family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320714.1 Intron NP_001307643.1
NM_005128.3 Intron NP_005119.2
XM_017028509.1 Intron XP_016883998.1

View Full Product Details