Product Details
- SNP ID
-
hCV26809471
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:150982256 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGTCCCTAAGAGAGGTCATCTGACT[G/T]GCTGTTCAGCCTAGGCTGCACACAC
- Phenotype
-
MIM: 603319
MIM: 606920
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ANXA9
PubMed Links
Gene Details
- Gene
- ANXA9
- Gene Name
- annexin A9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003568.2 |
234 |
UTR 5 |
|
|
NP_003559.2 |
- Gene
- CERS2
- Gene Name
- ceramide synthase 2
There are no transcripts associated with this gene.
View Full Product Details