Product Details
- SNP ID
-
rs4255791
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:28263483 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGTAACAGCAGCTCAGGCAAAGCAA[A/G]TTGAGAGCAACAGGTAGGCAGGAGC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SBK1
PubMed Links
Gene Details
- Gene
- SBK1
- Gene Name
- SH3 domain binding kinase 1
View Full Product Details