Product Details
- SNP ID
-
rs4258614
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:50668131 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAGCACACAGGGCTGAACACTCGA[C/G]TTGGTGACCAGGAAACCAGGGAGCA
- Phenotype
-
MIM: 613281
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC101927272
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78629804] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC101927272
- Gene Name
- uncharacterized LOC101927272
There are no transcripts associated with this gene.
- Gene
- SNX20
- Gene Name
- sorting nexin 20
View Full Product Details