Product Details

SNP ID
rs4790603
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4276000 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCCTGCATGTTACCAAGCCAGTCT[C/T]TGTCTCTTCCTTCTGTACCCTAATT
Phenotype
MIM: 601569
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UBE2G1 PubMed Links

Gene Details

Gene
UBE2G1
Gene Name
ubiquitin conjugating enzyme E2 G1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003342.4 Intron NP_003333.1

View Full Product Details