Product Details
- SNP ID
-
rs191351
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:43959076 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGTTTAAAGAAACTGGAAATAACA[C/G]GTGACAAATTATAGGATTGTGGGTG
- Phenotype
-
MIM: 600781
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PYY
PubMed Links
Gene Details
- Gene
- PYY
- Gene Name
- peptide YY
View Full Product Details