Product Details

SNP ID
rs197130
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:7988481 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGGATTTTCAACTGGCTAACCTTGG[C/T]TTCCACAACTGAATTTTTTTCAGAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BLOC1S5-TXNDC5 PubMed Links

Gene Details

Gene
BLOC1S5-TXNDC5
Gene Name
BLOC1S5-TXNDC5 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
PIP5K1P1
Gene Name
phosphatidylinositol-4-phosphate 5-kinase type 1 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details