Product Details

SNP ID
rs4871849
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:23107206 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAACAACAAAAAACCAAACAAACA[A/G]AAAAACAGAAAAACACAGAAAATTA
Phenotype
MIM: 603613
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC254896 PubMed Links

Gene Details

Gene
LOC254896
Gene Name
uncharacterized LOC254896
There are no transcripts associated with this gene.

Gene
TNFRSF10C
Gene Name
TNF receptor superfamily member 10c
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003841.3 Intron NP_003832.2

View Full Product Details