Product Details

SNP ID
rs2697366
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:156324877 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACAAACACTATGTGTCCCTTTTG[G/T]CAGGATTTTCTGATGTGTAATACTT
Phenotype
MIM: 601828
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NR4A2 PubMed Links

Gene Details

Gene
NR4A2
Gene Name
nuclear receptor subfamily 4 group A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006186.3 4795 UTR 3 NP_006177.1
XM_005246621.3 4795 UTR 3 XP_005246678.1
XM_005246622.3 4795 UTR 3 XP_005246679.1
XM_006712553.3 4795 UTR 3 XP_006712616.1
XM_011511246.1 4795 UTR 3 XP_011509548.1
XM_017004219.1 4795 UTR 3 XP_016859708.1
XM_017004220.1 4795 UTR 3 XP_016859709.1

View Full Product Details