Product Details

SNP ID
rs3180411
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:160435813 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGCCTACTTTAAAGTTGAGTGAT[A/G]GCATGAACTCTGAAAAGAGAATGCA
Phenotype
MIM: 605575 MIM: 616148
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SMC4 PubMed Links

Gene Details

Gene
SMC4
Gene Name
structural maintenance of chromosomes 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002800.2 3566 Intron NP_001002800.1
NM_001288753.1 3566 Intron NP_001275682.1
NM_005496.3 3566 Intron NP_005487.3
XM_006713459.3 3566 Intron XP_006713522.1
XM_011512311.2 3566 Intron XP_011510613.1
XM_011512312.2 3566 Intron XP_011510614.1
XM_017005491.1 3566 Intron XP_016860980.1
Gene
TRIM59
Gene Name
tripartite motif containing 59
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173084.2 3566 UTR 3 NP_775107.1

View Full Product Details