Product Details

SNP ID
rs3177959
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:73974671 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGCTACAAGAGAGGGAGGAGACG[C/T]CAAGGTTGAGCTTGCTTTATGGATG
Phenotype
MIM: 610263 MIM: 601693
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJB13 PubMed Links

Gene Details

Gene
DNAJB13
Gene Name
DnaJ heat shock protein family (Hsp40) member B13
There are no transcripts associated with this gene.

Gene
UCP2
Gene Name
uncoupling protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003355.2 1646 UTR 3 NP_003346.2

View Full Product Details