Product Details

SNP ID
rs3729715
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55154266 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATTTCCCGCACACCCAACTCCTC[C/T]ATCCTACACTCCTTTTTTATTCTCC
Phenotype
MIM: 614566 MIM: 191044 MIM: 191041
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAAF3 PubMed Links

Gene Details

Gene
DNAAF3
Gene Name
dynein axonemal assembly factor 3
There are no transcripts associated with this gene.

Gene
TNNI3
Gene Name
troponin I3, cardiac type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000363.4 Intron NP_000354.4
Gene
TNNT1
Gene Name
troponin T1, slow skeletal type
There are no transcripts associated with this gene.

View Full Product Details