Product Details

SNP ID
rs3765801
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:241532050 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
AGATGTTTTTGGACTTCAGATGAGA[A/C]GGGCTCCATTCATGGTTAATCATGA
Phenotype
MIM: 603538
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KMO PubMed Links

Gene Details

Gene
KMO
Gene Name
kynurenine 3-monooxygenase (kynurenine 3-hydroxylase)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003679.4 27 Intron NP_003670.2
XM_005273337.1 27 UTR 5 XP_005273394.1
XM_005273338.3 27 UTR 5 XP_005273395.1
XM_017002649.1 27 UTR 5 XP_016858138.1
XM_017002650.1 27 Intron XP_016858139.1

View Full Product Details