Product Details
- SNP ID
-
rs3739097
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:27137666 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCCGCAGGTCGTGTGCGATGCGGC[A/G]GTGCCCCAGCTGGACCACCAGGGCC
- Phenotype
-
MIM: 612195
MIM: 606395
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ABHD1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs567920330] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ABHD1
- Gene Name
- abhydrolase domain containing 1
There are no transcripts associated with this gene.
- Gene
- PREB
- Gene Name
- prolactin regulatory element binding
There are no transcripts associated with this gene.
- Gene
- PRR30
- Gene Name
- proline rich 30
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_178553.3 |
1244 |
Missense Mutation |
CGC,TGC |
R222C |
NP_848648.2 |
View Full Product Details